• USA Home
  • 569571-U - Discovery® Cyano Supelguard Cartridge

EMAIL THIS PAGE TO A FRIEND
569571-U Supelco

Discovery® Cyano Supelguard Cartridge

5 μm particle size, L × I.D. 2 cm × 3 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Cyano Supelguard Cartridge, Cyano Supelguard Guard Cartridge, Discovery Cyano, Discovery HPLC Columns,
description   Supelguard Cartridge
packaging   pkg of 2 ea
application(s)   HPLC: suitable
L × I.D.   2 cm × 3 mm
matrix active group   cyano phase
particle size   5 μm
pore size   180 Å
suitability   suitable for L10 per USP
separation technique   reversed phase
  reversed phase
  reversed phase

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Supelguard is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?