• USA Home
  • 577119-U - Titan C18 UHPLC Column, 1.9 micron

EMAIL THIS PAGE TO A FRIEND
577119-U Supelco

Titan C18 UHPLC Column, 1.9 micron

1.9 μm particle size, L × I.D. 15 cm × 2.1 mm

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, Titan C18 UHPLC Columns, 1.9 μm
Quality Level   100
feature   endcapped
mfr. no.   Titan
extent of labeling   13% carbon loading
parameter   18000 psi pressure (1241 bar)
  60 °C max. temp.
application(s)   UHPLC: suitable
L × I.D.   15 cm × 2.1 mm
matrix   monodisperse silica particle platform
matrix active group   C18 (octadecyl) phase
particle size   1.9 μm
pore size   0.76 cm3/g pore volume
  80 Å pore size
surface area   410 m2/g
pH range   2 - 8
suitability   suitable for L1 per USP
separation technique   reversed phase

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Titan is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Mycotoxin Analysis in Food and Feed: Detecting the Fungus Among Us

Mycotoxin production by various fungi on food and feed crops is an ever-present danger that requires regular and reliable analytical testing.  The ubiquitous presence of fungi in our fields, combined...
Keywords: Gas chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Metabolites, Separation, Solid phase extractions, Solvents

UHPLC Analysis of Taxanes Using A Titan C18 Column and UV Detection

Taxanes are diterpenes produced by the plants of the genus Taxus (yews), and are widely used as chemotherapy agents. Taxane agents include paclitaxel (Taxol) and docetaxel (Taxotere). The role of tax...
Keywords: Alzheimer Disease, Antitumor agents, Cancer, Chromatography, Clinical, Drug discovery, High performance liquid chromatography, Medicinal chemistry, Oncology, Pharmaceutical, Reductions, Separation

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?