• USA Home
  • 581330-U - Ascentis® C18 HPLC Column

EMAIL THIS PAGE TO A FRIEND
581330-U Supelco

Ascentis® C18 HPLC Column

5 μm particle size, L × I.D. 2 cm × 4.6 mm

  •  eCl@ss 32110501

Purchase

Properties

feature   endcapped
extent of labeling   25% Carbon loading
parameter   ≤70 °C temp. range
application(s)   HPLC: suitable
L × I.D.   2 cm × 4.6 mm
surface coverage   3.7 μmol/m2
impurities   <5 ppm metals
matrix   silica gel high purity, spherical
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   100 Å
surface area   450 m2/g
operating pH range   2 - 8
suitability   suitable for (Similar to Phenomenex Luna C18)
  suitable for L1 per USP
mode of chromatography   reversed phase

Description

Features and Benefits

• Excellent retention
• Symmetric peak shape
• High reproducibility
• Complete LC-MS compatibility

General description

The Ascentis family of columns is the fourth generation of HPLC column technology from Supelco scientists. Ascentis columns are bonded on high purity, 100 Angstrom silica including 3, 5, and 10 micron particle size. Columns are designed for small molecule applications and are scalable from micro columns (1.0 mm I.D.) to preparative dimensions (50 mm I.D.). The family includes C18, C8, Phenyl, Si and embedded polar group phase, RP-Amide.

Ascentis C18 is an extremely stable and reliable first choice HPLC column that gives symmetric peak shape and excellent retention even for difficult compounds.

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

guard cartridge

Product #

Description

Add to Cart

581372-U Ascentis® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
581373-U Ascentis® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm

related product

Product #

Description

Add to Cart

58278 Reversed Phase Test Mix, certified reference material, in methanol: water (3:2) (varied conc.)
59660-U Supelguard Guard Cartridge Holder, Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Milli-Q® Water Purification Solutions