• USA Home
  • 581596-U - Ascentis® Si Validation Pack
EMAIL THIS PAGE TO A FRIEND
581596-U Supelco
Ascentis® Si Validation Pack
5 μm particle size, L × I.D. 25 cm × 4.6 mm, Validation Pack, pkg of 3 ea
  •  eCl@ss 32110501
Purchase
Properties
Related Categories Analytical/Chromatography, Ascentis HPLC Columns, Ascentis Si, Ascentis Si Validation Pack, Chromatography,
packaging   pkg of 3 ea
L × I.D.   25 cm × 4.6 mm
matrix active group   silica phase
particle size   5 μm
mode of chromatography   Hydrophilic interaction (HILIC)
  Normal phase
Description
General description
An Ascentis Silica Validation Pack makes it easy to demonstrate method reproducibility on 3 different lots. The validation pack contains a kit with 3 columns - 1 from each of 3 lots of bonded phase. And with Ascentis Silica columns, you can be assured that all three columns will meet your expectations.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?