• USA Home
  • 58404 - Stainless Steel Tubing Kits
EMAIL THIS PAGE TO A FRIEND
58404 Supelco
Stainless Steel Tubing Kits
O.D. × I.D. 1/32 in. × 0.005 in.
  •  eCl@ss 32119290
Purchase
Properties
Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Stainless Steel Couplings, Unions, Tees, Tubing, Stainless Steel Tubing,
material   stainless steel
O.D. × I.D.   1/32 in. × 0.005 in.
Description
General description
Ready-to-use lengths. Each kit contains ten 5 cm and ten 10 cm pieces of 1/16 in. tubing.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Footnote
1/32 in. Stainless Steel Tubing Kit - 316 stainless steel, 1/32 in. O.D. × 0.005 in. I.D., for narrowbore HPLC. Two 6 in./15cm and two 12 in./30cm pieces.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Milli-Q® Water Purification Solutions
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?