EMAIL THIS PAGE TO A FRIEND
58464 Supelco
Collett
stainless steel
Synonym: Dynaseal Fittings
Purchase
Properties
Related Categories Analytical/Chromatography, Collet, Dynaseal Fittings, HPLC Accessories, HPLC/UHPLC,
material   stainless steel
packaging   pkg of 2 ea
fitting   1/16 in. Dynaseal ferrule for 1/16 in. O.D. tubing
Description
Features and Benefits
Dynaseal hand-tight fittings will seal against pressures to over 7000psi/490kg/cm². Each fitting consists of a reusable nut, collet, and ferrule. The polymer ferrule will not deform stainless steel seats and can be reused. Except for the fitting for Rheodyne valves, these fittings are compatible with all 1/16 in. female compression-type fittings now used on HPLC columns.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Dynaseal is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?