• USA Home
  • 59138C40 - SUPELCOSIL SAX1 HPLC Column

EMAIL THIS PAGE TO A FRIEND
59138C40 Supelco

SUPELCOSIL SAX1 HPLC Column

5 μm particle size, L × I.D. 25 cm × 4 mm

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, Ion Exchange Phases, SAX1,
Quality Level   100
mfr. no.   SUPELCOSIL
extent of labeling   12% carbon loading
parameter   0-70 °C temperature
  400 bar pressure (5801 psi)
application(s)   HPLC: suitable
L × I.D.   25 cm × 4 mm
matrix   silica gel, spherical particle platform
matrix active group   propyltrimethylammonium phase
particle size   5 μm
pore size   120 Å
surface area   170 m2/g
suitability   suitable for L14 per USP
Featured Industry   Food and Beverages
separation technique   anion exchange

Description

General description

The SUPELCOSIL SAX1 column has a strongly basic quaternary aminopropyl phase and is used for separating anions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
What is the functional group of the Supelcosil SAX1 HPLC column, Product 59138C40?
The SAX1 is packed with trimethylaminopropyl functionalized silica (5μm, 120Å), endcapped with TMCS. The ion exchange capacity is 0.4 to 0.5 meq/g.The SAX1 is used for the analysis of anionic organic compounds, such as glyphosate.Supelco does not recommend the SAX1 column for IC use.
What are the operating conditions for the Supelcosil SAX1 HPLC Column, Product 59138C40?
The SAX1 can be used with any HPLC mobile phase between pH 2-7.5.To control ionic retention, use pH and ionic strength to reduce retention.To control reverse phase retention, use acetonitrile to decrease retention.We do recommend avoiding methanol, and storing in acetonitrile, to prolong column life.
What guard do I use with the 4.0mm i.d. Supelcosil SAX1 column, Product 59138C40?
You can use the Supelguard SAX1Kit (59536-U), which contains one cartridge in a stand-alone holder with tubing & fittings to connect it in front of your Supelcosil SAX1 25cm x 4.0mm analytical column (59138C40). The replacement Supelguard SAX1 cartridges come as a 2-pack (59537-U).
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard cartridge

Product #

Description

Add to Cart

59536-U SUPELCOSIL SAX1 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?