• USA Home
  • 59182 - SUPELCOSIL LC-18 HPLC Column

EMAIL THIS PAGE TO A FRIEND
59182 Supelco

SUPELCOSIL LC-18 HPLC Column

12 μm particle size, L × I.D. 25 cm × 4.6 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical column, 12 micron, Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, LC-18,
Quality Level   100
feature   endcapped
mfr. no.   SUPELCOSIL
extent of labeling   11.0% carbon loading
parameter   ≤70 °C temp. limit
  400 bar pressure (5801 psi)
application(s)   HPLC: suitable
L × I.D.   25 cm × 4.6 mm
surface coverage   surface coverage 3.1 μmol/m2
matrix   silica gel, spherical particle platform
matrix active group   C18 (octadecyl) phase
particle size   12 μm
pore size   120 Å
surface area   170 m2/g
pH range   2 - 7.5
suitability   suitable for L1 per USP
Featured Industry   Food and Beverages
separation technique   reversed phase

Description

General description

A general purpose hydrophobic alkyl phase that is very retentive and gives good peak shape for a wide variety of compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

related product

Product #

Description

Add to Cart

58278 Reversed Phase Test Mix, certified reference material, in methanol: water (3:2) (varied conc.)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?