EMAIL THIS PAGE TO A FRIEND
59461

OPTI-SEAL pump seal

for use with 250, Series 4, 200, 400, 410, 620, Integral 4000, for use with Series 1, 2, 3, 3B, 10, PerkinElmer, 0990-7324, pkg of 1

Synonym: Optimize Technologies HPLC Pump Part for PerkinElmer®

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC Accessories, HPLC/UHPLC, Optimize Technologies Inc. Pump Replacement Parts, PerkinElmer Pump Replacement Parts,
packaging   pkg of 1
mfr. no.   PerkinElmer, 0990-7324
compatibility   for use with 250, Series 4, 200, 400, 410, 620, Integral 4000
  for use with Series 1, 2, 3, 3B, 10

Description

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?