• USA Home
  • 59604 - SUPELCOSIL ABZ+Plus Supelguard Cartridge

EMAIL THIS PAGE TO A FRIEND
59604 Supelco

SUPELCOSIL ABZ+Plus Supelguard Cartridge

5 μm particle size, L × I.D. 2 cm × 2.1 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories ABZ+Plus alkylamide, Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, Polar Bonded Phases,
packaging   pkg of 1 kit
application(s)   HPLC: suitable
L × I.D.   2 cm × 2.1 mm
matrix active group   amide, alkyl phase
particle size   5 μm
pore size   120 Å
suitability   suitable for L60 per USP
Featured Industry   Food and Beverages
separation technique   reversed phase

Description

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Supelguard is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?