• USA Home
  • 65621-U - Discovery® ZR Column Holder

EMAIL THIS PAGE TO A FRIEND
65621-U Supelco

Discovery® ZR Column Holder

for use with Discovery ZR Columns, pk of 1

  •  NACRES SB.52

Purchase

Properties

Quality Level   100
packaging   pk of 1
Featured Industry   Food and Beverages
compatibility   for use with Discovery ZR Columns

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?