• USA Home
  • 65805-U - Discovery® Zr-CarbonC18 Supelguard Cartridge

EMAIL THIS PAGE TO A FRIEND
65805-U Supelco

Discovery® Zr-CarbonC18 Supelguard Cartridge

5 μm particle size, L × I.D. 1 cm × 2.1 mm, pkg of 1 kit

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, C18 Supelguard Cartridge, Discovery HPLC Columns, Discovery Zirconia, Discovery Zr-Carbon C18,
Quality Level   100
description   Supelguard Cartridge
packaging   pkg of 1 kit
application(s)   HPLC: suitable
L × I.D.   1 cm × 2.1 mm
matrix   zirconia particle platform
matrix active group   octadecylphenyl phase
particle size   5 μm
pore size   300 Å
Featured Industry   Food and Beverages
separation technique   reversed phase

Description

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Supelguard is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

required but not provided

Product #

Description

Add to Cart

65621-U Discovery® ZR Column Holder, for use with Discovery ZR Columns, pk of 1

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?