• USA Home
  • 67201-U - BIOshell A160 Peptide C18, 5 μm HPLC Column

EMAIL THIS PAGE TO A FRIEND
67201-U Supelco

BIOshell A160 Peptide C18, 5 μm HPLC Column

5 μm particle size, L × I.D. 5 cm × 75 μm

Purchase

Properties

Related Categories Analytical/Chromatography, BIOshell A160 Peptide C18, 5 μm HPLC Columns, BIOshell HPLC Columns, C18 for Biopolymers, C18, 5 μm Capillary Columns,
material   stainless steel hardware (column)
description   Shell thickness (0.6 μm)
  Solid Core (3.5 μm)
feature   endcapped: no
extent of labeling   4.0% carbon loading
parameter   600 bar max. pressure
  90 °C max. temp.
application(s)   HPLC: suitable
L × I.D.   5 cm × 75 μm
surface coverage   2.2 μmol/m2
matrix   particle platform
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   160 Å
surface area   60 m2/g
operating pH range   1 - 8
suitability   suitable for L1 per USP
separation technique   reversed phase

Description

General description

BIOshell 160Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins. These columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected C18 ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90 °C). This greatly expands the application range for the separation of biomolecules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?