• USA Home
  • 67493-U - BIOshell A400 Protein C18 Column

EMAIL THIS PAGE TO A FRIEND
67493-U Supelco

BIOshell A400 Protein C18 Column

L × I.D. 15 cm × 100 μm, 3.4 μm particle size

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, BIOshell A400 Protein C18, 3.4 μm HPLC Columns, BIOshell HPLC Columns, C18 for Biopolymers, C18, 3.4 μm HPLC Columns,
Quality Level   100
material   stainless steel hardware (column)
description   Shell Thickness (0.2 μm)
  Solid Core (3.0 μm)
feature   endcapped
analyte chemical class(es)   proteins
mfr. no.   BIOshell
parameter   600 bar max. pressure
  90 °C max. temp.
application(s)   HPLC: suitable
L × I.D.   15 cm × 100 μm
matrix   spherical silica particle platform
matrix active group   C18 (octadecyl) phase
particle size   3.4 μm
pore size   3.4 μm particle size
  400 Å
surface area   ~15 m2/g
operating pH   1 - 9
suitability   suitable for HPLC
  suitable for L1 per USP
separation technique   reversed phase

Description

General description

BIOshell 400 Å Protein C18 columns contain 3.4 μm advanced Fused-Core® particles with 400 Å pores that are specifically engineered to provide efficient separation of larger proteins. These columns offer maximum hydrophobicity for resolving tricky hydrophilic and PEGylated proteins. The 3.4 μm Fused-Core particles allow a shorter analysis time yet similar back pressure to fully porous 1.7 μm, 300 Å fully porous C18 particles. These columns can be operated long term at pressures up to 9000 psi (600 bar), temperatures up to 90 °C, and within the pH range of 1 to 9.

Application

UHPLC Analysis of Proteins on BIOshell A160 Peptide C18 2.7 μm; BIOshell A400 Protein C18, 3.4 μm; and BIOshell IgG 1000 Å C18, 2.7 μm

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

related product

Product #

Description

Add to Cart

00922 Difluoroacetic acid, for LC-MS LiChropur, ≥97.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?