• USA Home
  • 73033ASTINT - Astec® Chiraldex® G-TA Intuvo Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
73033ASTINT Supelco

Astec® Chiraldex® G-TA Intuvo Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.12 μm, for use with Agilent Intuvo GC

Synonym: Astec® CHIRALDEX® G-TA Capillary GC Column

Purchase

Properties

material   fused silica
description   GC capillary column
df   0.12 μm
L × I.D.   30 m × 0.25 mm
matrix active group   non-bonded; 2,6-di-O-pentyl-3-trifluoroacetyl derivative of γ-cyclodextrin phase
column type   capillary chiral
compatibility   for use with Agilent Intuvo GC
separation technique   chiral

Description

General description

Astec® CHIRALDEX G-TA is the first choice in the Group 1 CSPs (Surface Interactions, Complex Derivatives). This phase has been shown to be the most broadly-selective phase for the pharmaceutical industry, especially for the analysis of chiral intermediates and drug studies in various stages of clinical trials. Separations occur without the inclusion mechanism and are typically faster and more efficient than most chiral stationary phases. G-TA has also been used to separate parent drug enantiomers and their metabolites. G-TA has its highest selectivity for oxygen-containing analytes like alcohols, diols and polyols as the free alcohol and as an acyl derivative; amines as acyl derivatives; amino alcohols, halogens (Cl>Br>F), amino acids, hydroxy acids, lactones, furans and pyrans. It is also highly selective for halogenated compounds.

Chem/Phys Resistance

Temp. Limits:
• -10 °C to 180 °C isothermal and programmed

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Gas Chromatography Columns for the Agilent® Intuvo GC System

Available now! Get your favorite Supelco® GC columns for use with the Agilent® Intuvo GC System!
Keywords: Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?