• USA Home
  • 77023ASTINT - Astec®CHIRALDEX® B-DM Intuvo Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
77023ASTINT Supelco

Astec®CHIRALDEX® B-DM Intuvo Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.12 μm, for use with Agilent (Intuvo GC)

Synonym: Astec® CHIRALDEX® B-DM Capillary GC Column

Purchase

Properties

material   fused silica
description   GC capillary column
df   0.12 μm
L × I.D.   30 m × 0.25 mm
matrix active group   non-bonded; 2,3-di-O-methyl-6-t-butyl silyl derivative of β-cyclodextrin phase
column type   capillary chiral
compatibility   for use with Agilent (Intuvo GC)
separation technique   chiral

Description

General description

Through special derivatization techniques, the concentration of the cyclodextrin in the CHIRALDEX B-DM has been substantially increased in the polysiloxane carrier. This phase is very useful for a number of free acids and bases. The B-DM is able to perform most of the separations done on a beta-permethylated phase, but with higher resolution. The selectivity of the B-DM covers applications of both the B-PM and B-PH phases, although with superior performance.

Chem/Phys Resistance

Temp. Limits:
• -10 °C to 200 °C isothermal, 220 °C programmed

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Gas Chromatography Columns for the Agilent® Intuvo GC System

Available now! Get your favorite Supelco® GC columns for use with the Agilent® Intuvo GC System!
Keywords: Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?