EMAIL THIS PAGE TO A FRIEND
78270 Supelco

1-Decene

New

reference material

  • CAS Number 872-05-9

  • Linear Formula CH3(CH2)7CH=CH2

  • Molecular Weight 140.27

  •  Beilstein/REAXYS Number 1737236

  •  MDL number MFCD00009577

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Extractables and Leachables, Food & Beverage Standards, Food Contact Materials,
grade   reference material
vapor density   4.84 (vs air)
vapor pressure   1.6 mmHg ( 23.8 °C)
assay   ≥97.0% (GC)
autoignition temp.   455 °F
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.421 (lit.)
bp   166.5-173.5 °C (lit.)
mp   −66.3-−66 °C (lit.)
density   0.741 g/mL at 25 °C (lit.)
storage temp.   −20°C
SMILES string   CCCCCCCCC=C
InChI   1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3
InChI key   AFFLGGQVNFXPEV-UHFFFAOYSA-N

Description

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
WGK Germany 
WGK 3
RTECS 
HE2071401
Flash Point(F) 
111.2 °F - closed cup
Flash Point(C) 
44.0 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?