• USA Home
  • 807648 - TSKgel® DEAE-SW Guardgel Kit

EMAIL THIS PAGE TO A FRIEND
807648

TSKgel® DEAE-SW Guardgel Kit

phase DEAE (diethylaminoethyl), L × I.D. 2.5 cm × 6 mm, 20 μm particle size

Synonym: TSKgel® DEAE-SW Columns

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, Silica-based TSKgel Anion Exchange Columns, TSKgel Anion Exchange HPLC Columns,
feature   counter ion H2PO4-
mfr. no.   Tosoh, 07648
parameter   0-100% organic solvent
  0.05-0.1 M salt concentration (starting buffer)
  0.2-0.5 M salt concentration (final buffer)
  10-45 °C temperature
L × I.D.   2.5 cm × 6 mm
matrix   silica particle platform
matrix active group   DEAE (diethylaminoethyl) phase
particle size   20 μm
operating pH   2.0 - 7.5
separation technique   anion exchange

Description

General description

TSKgel Guardgel kits contain:
• 5 mL Guardgel packing material
• Column holder, consisting of external reducing union, cap, frit, 2 ferrules
• Column, 25 mm x 9.5 mm OD x 6 mm ID, 2 μm frit at 10 mm depth, 1/16"" exit
• 2 Replacement frits
• Capillary tubing, 5 cm x 1/16"" OD x 0.01"" ID
• Nuts and ferrules, 10-32 threads for 1/16"" tubing, 2 each
• All hardware is 316 stainless steel

Dimensions for Guardgel kits for 21.5 mm ID columns:
• Column, 35 mm x 12.75 mm OD x 10 mm ID, 10 μm frit at 20mm depth, 1/16""
• Capillary tubing, 5 cm x 1/16"" OD x 0.02"" ID

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Central nervous system
WGK Germany 
WGK 2
Flash Point(F) 
55.4 °F
Flash Point(C) 
13 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard column

Product #

Description

Add to Cart

807168 TSKgel® DEAE-SW Columns, phase DEAE (diethylaminoethyl), L × I.D. 25 cm × 4.6 mm, 5 μm particle size
807163 TSKgel® DEAE-SW Columns, phase DEAE (diethylaminoethyl), L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?