• USA Home
  • 816133 - TSKgel® G1000Hxl for Light Scattering 30 cm x 7.8 mm ID, 5 μm

EMAIL THIS PAGE TO A FRIEND
816133

TSKgel® G1000Hxl for Light Scattering 30 cm x 7.8 mm ID, 5 μm

phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

Synonym: TSKgel® G1000HXL HPLC Column for Light Scattering Applications

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, TSKgel Gel Permeation Columns for Organic-Soluble Polymers, TSKgel HXL Type Gel Permeation Columns,
material   stainless steel
mfr. no.   Tosoh, 16133
parameter   1.0 mL/min flow rate
  50 bar max. pressure
  60 °C temperature
L × I.D.   30 cm × 7.8 mm
matrix   polystyrene-divinylbenzene
matrix active group   polystyrene-divinylbenzene phase
particle size   5 μm
suitability   suitable for L21 per USP
separation technique   size exclusion (SEC)

Description

General description

TSKgel® G1000Hxl columns are packed with rigid, porous, polymer beads. TSKgel G1000Hxl columns are applied to the non-aqueous size-exclusion separation of small molecules and oligomers less than 1,000 Da. As the number of solvents that can be exchanged directly with the shipping solvent (THF) is limited, use TSKgel HHR-type columns when a broader range of solvents is needed. The packing material in PN 816133 has been specially treated to remove sub-micron particles to aid in light scattering measurements.

Compatibility

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane

Physical form

Shipped in THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?