• USA Home
  • 818391 - TSKgel® GMHHR-H(30) HT HPLC Column

EMAIL THIS PAGE TO A FRIEND
818391

TSKgel® GMHHR-H(30) HT HPLC Column

phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 30 μm particle size

Synonym: TSKgel® GPC Columns for Applications up to 140° C

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, High Temp TSKgel Gel Permeation Columns (HT=140°C), TSKgel Gel Permeation Columns for Organic-Soluble Polymers,
material   stainless steel
mfr. no.   Tosoh, 18391
parameter   140 °C temperature
  15 bar max. pressure
  2.5 mL/min flow rate
application(s)   HPLC: suitable
L × I.D.   30 cm × 7.8 mm
matrix   polystyrene-divinylbenzene
matrix active group   polystyrene-divinylbenzene phase
particle size   30 μm
suitability   suitable for L21 per USP
separation technique   size exclusion (SEC)

Description

General description

TSKgel HT (High Temperature) columns are for the analysis of organic-soluble polymers at temperatures up to 140°C and are shipped in o-dichlorobenzene (ODCB), which can be exchanged for 1-chloronaphthalene and trichlorobenzene. The TSKgel mixed bed columns are packed with particles of different pore sizes that are blended to provide an extended linear calibration curve. Mixed bed columns are used when a broad range of polymer molar masses is expected in the sample. Shearing, which can be observed when analyzing ultra-high molar mass polymers on columns packed with 5 micron or smaller particles, can be overcome by using the TSKgel GMHhr-H(S) HT column, which is packed with 13 micron particles. The (S) is a reference to this shearing effect.

Physical form

ODCB can be exchanged by 1-chloronaphthalene and trichlorobenzene.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard column

Product #

Description

Add to Cart

818396 TSKgel® GMH<SUB>HR</SUB>-H (20) and (30) HT Guard column, phase polystyrene-divinylbenzene, L × I.D. 7.5 cm × 7.5 mm, 30 μm particle size

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?