• USA Home
  • 819007 - TSKgel® Phenyl-5PW RP Guard Cartridge

EMAIL THIS PAGE TO A FRIEND
819007

TSKgel® Phenyl-5PW RP Guard Cartridge

phase phenyl, L × I.D. 1.5 cm × 3.2 mm, 10 μm particle size, pkg of 3 ea

Synonym: TSKgel® Reversed Phase HPLC Guard Cartridge

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories, TSKgel Phenyl-5PW RP Columns (1000 Å pores),
material   stainless steel
packaging   pkg of 3 ea
mfr. no.   Tosoh Phenyl-5PW, 19007
extent of labeling   1 meq/mL ligand density (estimate)
parameter   no limitation salt concentration
  0-100% organic solvent
  4-60 °C temperature
application(s)   HPLC: suitable
L × I.D.   1.5 cm × 3.2 mm
matrix   polymer particle platform
matrix active group   phenyl phase
particle size   10 μm
pore size   1000 Å
operating pH   2 - 12
separation technique   reversed phase

Description

General description

Guard Cartridge for 4.6mmID TSKgel Phenyl-5PW RP columns, 3pk

TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.

Reversed-phase liquid chromatography (RPLC) is the preferred method for analysis and purification of polar and nonpolar compounds that are soluble in mixtures of water and organic solvents.

Physical form

Shipped in 30% methanol - 70% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

required but not provided

Product #

Description

Add to Cart

819018 Holder for 3.2 mm I.D. TSKgel® Guard Cartridges, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?