• USA Home
  • 820011 - TSKgel® VMpak-25 HPLC Column

EMAIL THIS PAGE TO A FRIEND
820011 Supelco

TSKgel® VMpak-25 HPLC Column

phase ammonium ion, quaternary, L × I.D. 5 cm × 2 mm, 7 μm particle size

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories, TSKgel Speciality Columns More...
material   stainless steel
mfr. no.   Tosoh, 20011
parameter   ≤0.5 M salt concentration
  0-100% organic solvent
  0.22 mL/min flow rate
  10-80 °C temperature
  20 bar max. pressure
application(s)   HPLC: suitable
L × I.D.   5 cm × 2 mm
matrix   hydroxylated methacrylic polymer
matrix active group   ammonium ion, quaternary phase
particle size   7 μm
operating pH   2 - 12
separation technique   mixed mode

Description

General description

TSKgel® VMpak-25 columns are specialty columns for the analysis of basic drugs that are often difficult to retain with good peak shape on alkyl-bonded reversed phase columns. The packing material in TSKgel VMpak-25 columns is a hydrophilic vinyl polymer gel (7 μm), similar to that in TSKgel Alpha-2500 size-exclusion columns, although the column dimensions are more appropriate for non-SEC applications. Depending on the mobile phase composition, compounds will be retained by a combination of reverse phase and cation exchange on residual carboxylic acid groups.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?