• USA Home
  • 821381 - ToyoScreen® PPG-600M Process Development Column

EMAIL THIS PAGE TO A FRIEND
821381 Supelco

ToyoScreen® PPG-600M Process Development Column

phase polypropylene glycol, pkg of 6 × 5 mL, 65 μm particle size

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories, ToyoScreen Hydrophobic Interaction Process Development Columns,
material   aluminum seal
  polypropylene and polyethylene frit
packaging   pkg of 6 × 5 mL
mfr. no.   Tosoh, 21381
parameter   ≤50% organic solvent
  1-5 mL/min flow rate
  20 mL/min max. flow rate
  3 bar max. pressure
application(s)   HPLC: suitable
L × I.D.   3 cm × 14.6 mm
matrix   hydroxylated methacrylic polymer
matrix active group   polypropylene glycol phase
particle size   65 μm
operating pH   1 - 13
capacity   55 g/L±10 adsorption capacity (lysozyme)
separation technique   hydrophobic interaction (HIC)

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Legal Information

ToyoScreen is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?