• USA Home
  • 822887 - TSKgel® GMHHR-H(30) HT2 HPLC Column

EMAIL THIS PAGE TO A FRIEND
822887

TSKgel® GMHHR-H(30) HT2 HPLC Column

phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 30 μm particle size

Synonym: TSKgel® GPC Columns for Applications up to 220° C

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, TSKgel Gel Permeation Columns for Organic-Soluble Polymers, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories,
material   stainless steel
mfr. no.   Tosoh, 22887
parameter   1 mL/min flow rate
  15 bar max. pressure
  220 °C temperature
application(s)   HPLC: suitable
L × I.D.   30 cm × 7.8 mm
matrix   polystyrene-divinylbenzene
matrix active group   polystyrene-divinylbenzene phase
particle size   30 μm
suitability   suitable for L21 per USP
separation technique   size exclusion (SEC)

Description

General description

TSKgel Ultra-High Temperature columns are for the analysis of organic-soluble polymers at extremely elevated temperatures. These columns are for applications up to 220°C and are shipped in o-dichlorobenzene (ODCB), which can be exchanged for 1-chloronaphthalene and trichlorobenzene.

The TSKgel mixed bed columns are packed with particles of different pore sizes that are blended to provide an extended linear calibration curve. Mixed bed columns are used when a broad range of polymer molar masses is expected in the sample.

Physical form

ODCB can be exchanged by 1-chloronaphthalene and trichlorobenzene.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

analytical column

Product #

Description

Add to Cart

822888 TSKgel® GMHHR-H(20) HT2 HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 20 μm particle size

guard column

Product #

Description

Add to Cart

822891 TSKgel® GMHHR-H (20) and (30) HT2 Guard Column, phase polystyrene-divinylbenzene, L × I.D. 7.5 cm × 7.5 mm, 30 μm particle size

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?