• USA Home
  • 842154 - TSKgel® DEAE-SW Guardgel Kit

EMAIL THIS PAGE TO A FRIEND
842154

TSKgel® DEAE-SW Guardgel Kit

phase DEAE (diethylaminoethyl), L × I.D. 1 cm × 2 mm, 5 μm particle size, pkg of 3 ea

Synonym: TSKgel® DEAE-SW Columns

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC & FPLC Columns for Biopolymer Separations, HPLC/UHPLC, Silica-based TSKgel Anion Exchange Columns, TSKgel Anion Exchange HPLC Columns,
feature   counter ion H2PO4-
packaging   pkg of 3 ea
mfr. no.   Tosoh, 42154
parameter   0-100% organic solvent
  10-45 °C temperature
application(s)   HPLC: suitable
L × I.D.   1 cm × 2 mm
matrix   silica particle platform
matrix active group   DEAE (diethylaminoethyl) phase
particle size   5 μm
pore size   125 Å
operating pH   2.0 - 7.5
separation technique   anion exchange

Description

General description

Guard cartridge for 2mmID TSKgel DEAE-2SW column, 3pk

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?