EMAIL THIS PAGE TO A FRIEND
861257 Supelco

Oxygenates Mix 1

certified reference material, in methanol (varied)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Biofuels/Fuels, Certified Reference Materials (CRMs),
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   in methanol (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Petroleum
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

tert-Amyl methyl ether 2000 μg/mL    
tert-Butanol 20000 μg/mL    
tert-Butyl ethyl ether 2000 μg/mL    
tert-Butyl methyl ether 2000 μg/mL    
Diisopropyl ether 2000 μg/mL    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?