• USA Home
  • 97763-U - VersaPak® Silica Cartridge

EMAIL THIS PAGE TO A FRIEND
97763-U Supelco

VersaPak® Silica Cartridge

20-45 μm, matrix active group Spherical Silica, I.D. × L 80 mm × 150 mm, pkg of 2

Purchase

Properties

Related Categories Chromatography, Labware, VersaFlash More...
composition   bed wt., 385 g
packaging   pkg of 2
parameter   60 psi max. pressure
I.D. × L   80 mm × 150 mm
matrix active group   Spherical Silica
particle size   20-45 μm

Description

Legal Information

VersaPak is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Milli-Q® Water Purification Solutions