• USA Home
  • MET260A - cis-1,2,3,6-Tetrahydrophthalimide

EMAIL THIS PAGE TO A FRIEND
MET260A Supelco

cis-1,2,3,6-Tetrahydrophthalimide

analytical standard

Synonym: cis-4-Cyclohexene-1,2-dicarboximide

  • CAS Number 1469-48-3

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein/REAXYS Number 82338

  •  EC Number 215-999-0

  •  MDL number MFCD00005880

  •  PubChem Substance ID 24897381

Purchase

Properties

grade   analytical standard
InChI Key   CIFFBTOJCKSRJY-OLQVQODUSA-N
form   neat
packaging   ampule of 5000 mg
mfr. no.   Chem Service, Inc., MET-260A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   129-133 °C (lit.)
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
352.4 °F
Flash Point(C) 
178 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?