EMAIL THIS PAGE TO A FRIEND
MET380B Supelco

Atrazine-desethyl

analytical standard

Synonym: 2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine, Desethylatrazine

  • CAS Number 6190-65-4

  • Empirical Formula (Hill Notation) C6H10ClN5

  • Molecular Weight 187.63

  •  Beilstein/REAXYS Number 610488

  •  MDL number MFCD00078646

  •  PubChem Substance ID 24897382

Purchase

Properties

grade   analytical standard
InChI Key   DFWFIQKMSFGDCQ-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., MET-380B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
XY5850570

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

36629 Atrazine-desethyl, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?