• USA Home
  • MET380D - Atrazine-desethyl-2-hydroxy

EMAIL THIS PAGE TO A FRIEND
MET380D Supelco

Atrazine-desethyl-2-hydroxy

analytical standard

  • CAS Number 19988-24-0

  • Empirical Formula (Hill Notation) C6H11N5O

  • Molecular Weight 169.18

  •  Beilstein/REAXYS Number 2974745

  •  MDL number MFCD00155384

  •  PubChem Substance ID 24897383

  •  NACRES NA.24

Purchase

Properties

grade   analytical standard
InChI Key   GCKLGRUZDXSATG-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., MET-380D
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

45490 Atrazine-desethyl-2-hydroxy, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?