• USA Home
  • MET380E - Atrazine-desethyl-desisopropyl-2-hydroxy

EMAIL THIS PAGE TO A FRIEND
MET380E Supelco

Atrazine-desethyl-desisopropyl-2-hydroxy

analytical standard

Synonym: 4,6-Diamino-2-hydroxy-1,3,5-triazine, Ammeline

  • CAS Number 645-92-1

  • Empirical Formula (Hill Notation) C3H5N5O

  • Molecular Weight 127.10

  •  Beilstein/REAXYS Number 607753

  •  EC Number 211-455-1

  •  MDL number MFCD00023186

  •  PubChem Substance ID 24897384

Purchase

Properties

grade   analytical standard
InChI Key   MASBWURJQFFLOO-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., MET-380E
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

45613 Atrazine-desethyl-desisopropyl-2-hydroxy, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?