• USA Home
  • MET40A - 4-Chloro-2-methylphenol

EMAIL THIS PAGE TO A FRIEND
MET40A Supelco

4-Chloro-2-methylphenol

analytical standard

Synonym: 4-Chloro-o-cresol

Purchase

Properties

grade   analytical standard
InChI Key   RHPUJHQBPORFGV-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., MET-40A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   220-225 °C(lit.)
mp   43-46 °C (lit.)
Featured Industry   Agriculture
format   neat
Gene Information   rat ... Ar(24208)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3437 - class 6.1 - PG 2 - EHS - Chlorocresols, solid
WGK Germany 
2
RTECS 
GO7120000
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?