• USA Home
  • MET58A - Atrazine-desisopropyl

EMAIL THIS PAGE TO A FRIEND
MET58A Supelco

Atrazine-desisopropyl

analytical standard

Synonym: 2-Amino-4-chloro-6-ethylamino-1,3,5-triazine

  • CAS Number 1007-28-9

  • Empirical Formula (Hill Notation) C5H8ClN5

  • Molecular Weight 173.60

  •  Beilstein Registry Number 8150

  •  MDL number MFCD00078645

  •  PubChem Substance ID 24897390

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Atrazine metabolites,
grade   analytical standard
InChI Key   IVENSCMCQBJAKW-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., MET-58A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?