EMAIL THIS PAGE TO A FRIEND
MET652A Supelco

Disulfoton-sulfone

analytical standard

  • CAS Number 2497-06-5

  • Empirical Formula (Hill Notation) C8H19O4PS3

  • Molecular Weight 306.40

  •  Beilstein Registry Number 1713538

  •  MDL number MFCD00155401

  •  PubChem Substance ID 24897393

Purchase

Properties

grade   analytical standard
InChI Key   BKVJOVPVLOJPKJ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., MET-652A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3278 6.1 / PGII
WGK Germany 
3
RTECS 
TD8925000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?