• USA Home
  • MET652B - Disulfoton-sulfoxide

EMAIL THIS PAGE TO A FRIEND
MET652B Supelco

Disulfoton-sulfoxide

analytical standard

  • CAS Number 2497-07-6

  • Empirical Formula (Hill Notation) C8H19O3PS3

  • Molecular Weight 290.40

  •  Beilstein/REAXYS Number 1712528

  •  EC Number 219-679-1

  •  MDL number MFCD00155402

  •  PubChem Substance ID 24897394

  •  NACRES NA.24

Purchase

Properties

grade   analytical standard
InChI Key   UPUGLJYNCXXUQV-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., MET-652B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

General description

Disulfoton-sulfoxide is a transformation product of the organophosphorus pesticide disulfoton.

Application

Disulfoton-sulfoxide may be used as a reference standard for the determination of disulfoton-sulfoxide in water samples by automated on-line solid-phase extraction followed by thermospray liquid chromatography-mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
3
RTECS 
TD8600000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?