EMAIL THIS PAGE TO A FRIEND
MET81B Supelco

Endosulfan ether

ampule of 50 mg

  • CAS Number 3369-52-6

  • Empirical Formula (Hill Notation) C9H6Cl6O

  • Molecular Weight 342.86

  •  Beilstein/REAXYS Number 22311

  •  MDL number MFCD00168052

  •  PubChem Substance ID 24897400

  •  NACRES NA.24

Purchase

Properties

InChI Key   CKPWHXBGVRURFU-NYQGKTAPSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., MET-81B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
PC1224500
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?