EMAIL THIS PAGE TO A FRIEND
PS1012

Fluchloralin

analytical standard

  • CAS Number 33245-39-5

  • Empirical Formula (Hill Notation) C12H13ClF3N3O4

  • Molecular Weight 355.70

  •  Beilstein/REAXYS Number 2778716

  •  EC Number 251-426-0

  •  MDL number MFCD00078686

  •  PubChem Substance ID 24899075

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-1012
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   CCCN(CCCl)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
InChI   1S/C12H13ClF3N3O4/c1-2-4-17(5-3-13)11-9(18(20)21)6-8(12(14,15)16)7-10(11)19(22)23/h6-7H,2-5H2,1H3
InChI key   MNFMIVVPXOGUMX-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
XU5160000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?