EMAIL THIS PAGE TO A FRIEND
PS1015 Supelco

Desmedipham

analytical standard

  • CAS Number 13684-56-5

  • Empirical Formula (Hill Notation) C16H16N2O4

  • Molecular Weight 300.31

  •  Beilstein Registry Number 2395716

  •  EC Number 237-198-5

  •  MDL number MFCD00055462

  •  PubChem Substance ID 24899077

Purchase

Properties

grade   analytical standard
InChI Key   WZJZMXBKUWKXTQ-UHFFFAOYSA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1015
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
FD0425000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?