EMAIL THIS PAGE TO A FRIEND
PS1026 Supelco

Mephosfolan

analytical standard

Synonym: Diethyl (4-methyl-1,3-dithiolan-2-ylidene)phosphoramidate

Purchase

Properties

grade   analytical standard
InChI Key   LTQSAUHRSCMPLD-CMDGGOBGSA-N
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-10265
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
3
RTECS 
JP1050000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?