• USA Home
  • PS1030 - δ-(cis-trans)-Phenothrin

EMAIL THIS PAGE TO A FRIEND
PS1030

δ-(cis-trans)-Phenothrin

analytical standard

Synonym: Phenothrin

  • CAS Number 26002-80-2

  • Empirical Formula (Hill Notation) C23H26O3

  • Molecular Weight 350.45

  •  Beilstein/REAXYS Number 8788185

  •  EC Number 247-404-5

  •  MDL number MFCD00078716

  •  PubChem Substance ID 24899087

Purchase

Properties

grade   analytical standard
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-1030
Featured Industry   Agriculture
Environmental
SMILES string   C\C(C)=C\C1C(C(=O)OCc2cccc(Oc3ccccc3)c2)C1(C)C
InChI   1S/C23H26O3/c1-16(2)13-20-21(23(20,3)4)22(24)25-15-17-9-8-12-19(14-17)26-18-10-6-5-7-11-18/h5-14,20-21H,15H2,1-4H3
InChI key   SBNFWQZLDJGRLK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?