EMAIL THIS PAGE TO A FRIEND
PS1040 Supelco

Methoprene

analytical standard

  • CAS Number 40596-69-8

  • Empirical Formula (Hill Notation) C19H34O3

  • Molecular Weight 310.47

  •  Beilstein/REAXYS Number 1913191

  •  EC Number 254-993-2

  •  MDL number MFCD00072475

  •  PubChem Substance ID 24899092

Purchase

Properties

grade   analytical standard
InChI Key   NFGXHKASABOEEW-LDRANXPESA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1040
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
JR1685000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?