EMAIL THIS PAGE TO A FRIEND
PS1045 Supelco

Ethofumesate

analytical standard

  • CAS Number 26225-79-6

  • Empirical Formula (Hill Notation) C13H18O5S

  • Molecular Weight 286.34

  •  Beilstein Registry Number 5759730

  •  EC Number 247-525-3

  •  MDL number MFCD00055321

  •  PubChem Substance ID 24899094

Purchase

Properties

grade   analytical standard
InChI Key   IRCMYGHHKLLGHV-UHFFFAOYSA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1045
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
DF7716500
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?