EMAIL THIS PAGE TO A FRIEND
PS1060 Supelco

Fenbutatin oxide

analytical standard

Synonym: Bis[tris-(2-methyl-2-phenylpropyl)tin] oxide

Purchase

Properties

grade   analytical standard
InChI Key   HOXINJBQVZWYGZ-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-1060
Featured Industry   Agriculture
Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - SMP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
JN8770000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?