EMAIL THIS PAGE TO A FRIEND
PS1065 Supelco

Chlorosulfuron

analytical standard

Synonym: 1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea, Chlorsulfuron

  • CAS Number 64902-72-3

  • Empirical Formula (Hill Notation) C12H12ClN5O4S

  • Molecular Weight 357.77

  •  Beilstein/REAXYS Number 577255

  •  EC Number 265-268-5

  •  MDL number MFCD00128059

  •  PubChem Substance ID 24899106

Purchase

Properties

grade   analytical standard
InChI Key   VJYIFXVZLXQVHO-UHFFFAOYSA-N
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1065
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
YS6640000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?