EMAIL THIS PAGE TO A FRIEND
PS1067

Cymoxanil

analytical standard

Synonym: 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea

  • CAS Number 57966-95-7

  • Empirical Formula (Hill Notation) C7H10N4O3

  • Molecular Weight 198.18

  •  Beilstein/REAXYS Number 2214018

  •  EC Number 261-043-0

  •  MDL number MFCD00137381

  •  PubChem Substance ID 24899107

Purchase

Properties

grade   analytical standard
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1067
Featured Industry   Agriculture
Environmental
SMILES string   CCNC(=O)NC(=O)C(=N\OC)\C#N
InChI   1S/C7H10N4O3/c1-3-9-7(13)10-6(12)5(4-8)11-14-2/h3H2,1-2H3,(H2,9,10,12,13)/b11-5+
InChI key   XERJKGMBORTKEO-VZUCSPMQSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, thymus
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
AB5957000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?