EMAIL THIS PAGE TO A FRIEND
PS1071 Supelco

Mavrik

analytical standard

Synonym: τ-Fluvalinate, N-[2-Chloro-4-(trifluoromethyl)phenyl]-D-valine (RS)-cyano(3-phenoxyphenyl)methyl ester

  • CAS Number 102851-06-9

  • Empirical Formula (Hill Notation) C26H22ClF3N2O3

  • Molecular Weight 502.91

  •  Beilstein Registry Number 8092907

  •  MDL number MFCD00871307

  •  PubChem Substance ID 24899109

Purchase

Properties

grade   analytical standard
InChI Key   INISTDXBRIBGOC-XMMISQBUSA-N
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1071
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?