EMAIL THIS PAGE TO A FRIEND
PS1075 Supelco

Tilt

analytical standard

Synonym: Propiconazole

  • CAS Number 60207-90-1

  • Empirical Formula (Hill Notation) C15H17Cl2N3O2

  • Molecular Weight 342.22

  •  Beilstein/REAXYS Number 9349305

  •  EC Number 262-104-4

  •  MDL number MFCD00055299

  •  PubChem Substance ID 24899111

Purchase

Properties

grade   analytical standard
InChI Key   STJLVHWMYQXCPB-UHFFFAOYSA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1075
Featured Industry   Agriculture
Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XZ4620000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?