• USA Home
  • PS1102 - Dicamba methyl ester

EMAIL THIS PAGE TO A FRIEND
PS1102 Supelco

Dicamba methyl ester

analytical standard

Synonym: Methyl 3,6-dichloro-2-methoxybenzoate

  • CAS Number 6597-78-0

  • Empirical Formula (Hill Notation) C9H8Cl2O3

  • Molecular Weight 235.06

  •  MDL number MFCD00056244

  •  PubChem Substance ID 24899124

Purchase

Properties

grade   analytical standard
InChI Key   AWSBKDYHGOOSML-UHFFFAOYSA-N
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1102
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?