• USA Home
  • PS1106 - Mecoprop methyl ester

EMAIL THIS PAGE TO A FRIEND
PS1106

Mecoprop methyl ester

analytical standard

Synonym: Methyl 2-(4-chloro-2-methylphenoxy)propioate

  • CAS Number 2786-19-8

  • Empirical Formula (Hill Notation) C11H13ClO3

  • Molecular Weight 228.67

  •  Beilstein/REAXYS Number 2455097

  •  MDL number MFCD00055258

  •  PubChem Substance ID 329823643

Purchase

Properties

Related Categories All Herbicide Standards, Analytical Standards, Analytical/Chromatography, E-M, Herbicides,
grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1106
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   COC(=O)C(C)Oc1ccc(Cl)cc1C
InChI   1S/C11H13ClO3/c1-7-6-9(12)4-5-10(7)15-8(2)11(13)14-3/h4-6,8H,1-3H3
InChI key   YWGAULPFWIQKRB-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?