EMAIL THIS PAGE TO A FRIEND
PS1211 Supelco

Flumetralin

100 μg/mL in acetone, analytical standard

  • CAS Number 62924-70-3

  • Empirical Formula (Hill Notation) C16H12ClF4N3O4

  • Molecular Weight 421.73

  •  Beilstein/REAXYS Number 2956801

  •  MDL number MFCD00078695

  •  PubChem Substance ID 24898934

Purchase

Properties

grade   analytical standard
InChI Key   PWNAWOCHVWERAR-UHFFFAOYSA-N
packaging   ampule of 1 mL
mfr. no.   Chem Service, Inc., PS-1211
concentration   100 μg/mL in acetone
Featured Industry   Agriculture
Safety & Documentation

Safety Information

RIDADR 
UN 1090 3/PG 2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?